Catalogue login
Username: 
Password:
Login
New User
Register here
Forgotten password?
English website
  PRODUKTSUCHE
A question ?  
  We will
help you!
Do you have questions ?
Tel:+380 44 494 42 42
Fax:+380 44 494 42 42
info@hlr.com.ua
ХИМЛАБОРРЕАКТИВ ООО
ул. Щелковская, 8
07400 Бровары, Киевская обл.
Tel:+380 44 494 42 42
Fax:+380 44 494 42 42
Technical Service
Life Science
Tel:+380 44 593-77-70
Minizentrifuge   
Fisherbrand® Mini-Centrifuge 
An economic small-sized benchtop centrifuge suitable for many applications including quick spindowns from walls or caps of centrifuge tubes, microfiltration of HPLC samples and cell separations, and short-term coldroom use. The mini centrifuge is supplied with interchangeable rotors for microtubes and strip tubes.
    • Lid-activated ON/OFF switch delivers fast acceleration and deceleration.
    • Max. speed 6000-1
    • Max. force 2000 G
    • Standard rotor: 6 x 1.5/2.0 ml tubes
      (6 x 0.4/0.2 ml tubes with supplied adapter)
    • PCR Strip rotor: two 8 x 0.2 ml PCR tube strips or 16 x 0.2 ml tubes
    • Clear polycarbonate lid allows maximum visibility
    • Centrifuge base has nonskid rubber feet Ø/H: 140/110 mm
    • Dimensions:
Scope of delivery: Mini Centrifuge including Standard rotor, PCR strip rotor and adapters to spin 0.4 and 0.5 ml tubes.
Fisherbrand® Mini Centrifuge
Type
PU
pcs.
Item No.
EUR
Fisherbrand Mini Centrifuge including 2 rotors and adapters 1 3153793 Price Info
Six-place Standard rotor (6 x 0.2/0.5 ml) 1 3153788
PCR strip rotor (8 x 0.2 ml or 16 x 0.2 ml) 1 3153789
Adapters for 0.5/0.6 ml tubes 6 3153790
Adapters for 0.4/0.25 ml tubes 6 3153791
Adapters for 0.2 ml tubes 6 3153792
ХИМЛАБОРРЕАКТИВ ООО - ул. Щелковская, 8 - 07400 Бровары, Киевская обл. - Tel +380 44 494 42 42 - Fax. +380 44 494 42 42 - info@hlr.com.ua
Copyright © 2011 Thermo Fisher Scientific Inc.