Fisherbrand® Crimp Neck Vials ND8 and Seals Crimp
Neck Vials ND8 in different design and with different volumes available;
for e.g. CTC A200S, Beckman, Carlo Erba, Dynatech, Fisons, Gilson,
Agilent, VWR (Merck®)/Hitachi, Perkin Elmer, Shimadzu, Spark, Unicam, etc.
Vials that carry a CleanPack label on the front side of the PP-Box have
been packed in a certified cleanroom class 10,000. 8mm
Aluminium-Crimp Caps are available with different
liners: · Natural Rubber is ideal for multiple injections due to high resealability ·
Silicone
is the cleanest synthetic elastomer and is therefore used for critical
analysis.Temperature resistant from -60 °C up to 200 °C
·
Silicone
liners with PTFE on both sides for less coring Further seals upon request. |
![]() |
Fisherbrand® Crimp Neck Vials ND 8 | |||||||
Type | Capacity ml |
Size mm |
PU / pcs | Item No. | EUR | ||
Clear glass, flat bottom | 0.8 | 30x8.2 | 100 | 3205407 | Price Info | ||
Clear glass, flat bottom | 0.7 | 40x7 | 10x100 | 3205409 | |||
Amberglass, flat bottom | 0.7 | 40x7 | 10x100 | 3205410 | |||
Clear glass, rounded bottom | 0.3 | 31.5x5.5 | 10x100 | 3205406 | |||
Clear glass, conical | 0.3 | 31.5x5.5 | 10x100 | 3205411 | |||
Clear glass, flat bottom | 1.2 | 40x8.2 | 100 | 3205412 | |||
Amberglass, flat bottom | 1.2 | 40x8.2 | 100 | 3205413 | |||
Clear glass, conical | 0.7 | 40x7 | 10x100 | 3205408 | |||
Amber glass, conical | 0.7 | 40x7 | 10x100 | 3205405 | |||
Amber glass, conical | 0.4 | 30x7 | 10x100 | 3146109 |
![]() |
Fisherbrand® Aluminium Crimp Caps 8mm,
with Septa 8mm
Aluminium-Crimp Caps are available with different
liners: ·
Natural
Rubber is ideal for multiple injections due to high
resealability ·
Silicone
is the cleanest synthetic elastomer and is therefore used for critical
analysis. Temperature resistant from -60 °C up to 200 °C
·
Silicone
liners with PTFE on both sides for less coring |
Fisherbrand® Aluminium Crimp Caps 8mm, with Septa | |||||||
Type | Thickness mm |
Hardness | PU / pcs | Item No. | EUR | ||
Natural rubber / Butyl / TEF | 1.0 | 45° shore A | 100 | 3205140 | Price Info | ||
Natural rubber / TEF (Agilent Quality) | 1.0 | 60° shore A | 100 | 3205142 | |||
Silicone white / PTFE red UltraClean™ | 1.3 | 45° shore A | 100 | 3205141 | |||
PTFE red / Silicone white / PTFE red | 1.0 | 45° shore A | 100 | 3205139 | |||
Silicone white / PTFE red, slitted | 1.3 | 45° shore A | 100 | 3205143 |
Other Fisherbrand® Combination Seals for Crimp Neck Vials ND8
|
|
Other Fisherbrand® Combination Seals for Crimp Neck Vials ND8 | |||||||
Type | Septa Materials | Hardness | PU / pcs | Item No. | EUR | ||
PE Push-On Kappe, blue | with thinned penetration point | 100 | 3146124 | Price Info | |||
PE-Cap, clear, 9 x 5.9mm, 4mm hole |
Natural rubber red-orange / TEF clear 1.0mm | 60° shore A | 100 | 3146125 | |||
PE-Cap, clear, 9 x 5.9mm, 4mm hole | Silicone white/PTFE red 1.3mm | 45° shore A | 100 | 3146126 |
|
Fisherbrand® Crimp Neck
Vials ND11,
|
Fisherbrand® Crimp Neck Vials ND11 | |||||||
Type |
Capacity |
Size mm |
PU / pcs | Item No. | EUR | ||
Clear glass, with narrow opening | 1.5 | 32x11.6 | 100 | 3205455 | Price Info | ||
Clear glass, with wide opening | 1.5 | 32x11.6 | 100 | 3125619 | |||
Clear glass, with wide opening, label and graduation lines | 1.5 | 32x11.6 | 100 | 3205459 | |||
Amber glass, with wide opening, label and graduation lines | 1.5 | 32x11.6 | 100 | 3205460 | |||
Microliter vial, with 15µl inner cone | 1.1 | 32x11.6 | 100 | 3124595 | |||
Crimp neck micro vial, clear glass, conical | 1.1 | 32x11.6 | 100 | 3205456 | |||
Crimp neck vial with integrated micro insert | 0.2 | 32x11.6 | 100 | 3205463 | |||
TopSert, TPX vial, clear, with integrated glass micro insert | 0.2 | 32x11.6 | 100 | 3124628 | |||
TopSert, TPX vial, amber, with integrated glass micro insert | 0.2 | 32x11.6 | 100 | 3146110 |
Fisherbrand® Micro Insert for Vials with wide Opening | |||||||
Type |
Capacity |
Size mm |
PU / pcs | Item No. | EUR | ||
Clear glass, conical top, 15mm | 0.1 | 31x6 | 10x100 | 3205119 | Price Info | ||
Clear glass, with plastic spring | 0.1 | 29x5.7 | 10x100 | 3205125 | |||
Clear glass, with flat bottom | 0.2 | 31x6 | 10x100 | 3205136 | |||
Clear glass, conical top, 12mm | 0.1 | 31x6 | 10x100 | 3205127 |
Fisherbrand® Aluminium Bördelkappen 11mm, mit Septum
|
|
Fisherbrand® Aluminium Crimp Caps 11mm, with Septa | |||||||
Type | Thickness mm |
Hardness | PU / pcs | Item No. | EUR | ||
PTFE virginal | 0.25 | 53° shore D | 100 | 3205451 | Price Info | ||
Natural rubber red-orange / Butyl / TEF | 1.0 | 45° shore A | 100 | 3205448 | |||
Natural rubber red-orange / TEF (Agilent Quality) | 1.0 | 60° shore A | 100 | 3125620 | |||
Natural rubber red-orange / Butyl / TEF | 1.3 | 45° shore A | 100 | 3205454 | |||
PTFE grey / Butyl red / PTFE grey | 1.3 | 55° shore A | 100 | 3205453 | |||
Silicone white / PTFE red / UltraClean™ | 1.3 | 45° shore A | 100 | 3205447 | |||
PTFE red / Silicone white / PTFE red | 1.0 | 45° shore A | 100 | 3205446 | |||
Silicone white / PTFE blue, cross-slitted | 1.5 | 55° shore A | 100 | 3205452 |
![]() |
Fisherbrand® Magnetic Crimp Caps 11mm, with
Septa
|
Fisherbrand® Magnetic Crimp Caps 11mm, with Septa | |||||||
Type | Thickness mm |
Hardness | PU / pcs | Item No. | EUR | ||
Silicone white / PTFE red UltraClean™ | 1.3 | 45° shore A | 100 | 3205449 | Price Info | ||
PTFE red / Silicone white / PTFE red | 1.0 | 45° shore A | 100 | 3205450 |
Other Fisherbrand® Combination Seals for Crimp Neck
Vials ND11
|
![]() |
Other Fisherbrand® Combination Seals for Crimp Neck Vials ND11: PE-Caps, clear | |||||||
Type | Septa Material | Thickness mm |
Hardness | PU / pcs | Item No. | EUR | |
PE Push-On Cap, blue transparent | With thinned penetration point |
100 |
3146112 | Price Info | |||
PE-Cap, transparent, 13x7.5mm, 4.5mm hole | Natural rubber red-orange / TEF transparent | 1.0 | 60° shore A | 100 | 3205505 | ||
PE-Cap, transparent, 13x7.5mm, 4.5mm hole | Silicone white / PTFE red | 1.3 | 55° shore A | 100 | 3205506 |
![]() |
Fisherbrand® Headspace
Vials ND20 and Seals Our Headspace Vials are made out of glass of the
1st hydrolytic glass and differ regarding volume, type
and length of the neck, design of the bottom. For the choice of the correct vials we specified some
Autosampler -Compatibilities. Beside the correct choice of the septa it is
also important to choose the correct crimp cap. Headspace Caps are safety caps for Headspace analysis, which should avoid explosion of the vial in case of too much internal pressure. The Headspace Caps have special score-lines with bridges that break open at an inner pressure of 3.0 ± 0.5 bar. Thus the excess pressure escapes and the risk of vial explosion is avoided. Thus they correspond to the patented PERKIN ELMER Pressure Release Seal consisting of an Aluminium Cap with a slit, a metal star washer plate and a liner with ears. This system only releases excess pressure reliably when using a vial with such a top. |
Fisherbrand® Headspace Vials ND20 | |||||||
Type |
Capacity |
Size mm |
PU / pcs | Item No. | EUR | ||
Clear glass, HS-Crimp Neck, rounded bottom (Perkin Elmer) |
5 | 38.25x22 | 100 | 3205095 | Price Info | ||
Clear glass, DIN-Crimp Neck, rounded bottom (Carlo Erba, CTC, Fisons, Varian, CP) |
10 | 46x22.5 | 100 | 3205554 | |||
Amber glass, DIN-Crimp Neck, rounded bottom (Carlo Erba, CTC, Fisons, Varian, CP) |
10 | 46x22.5 | 100 | 3146113 | |||
Clear glass, DIN-Crimp Neck, flat bottom (Carlo Erba, Dani, Fisons, Agilent) |
10 | 46x22.5 | 100 | 3205550 | |||
Clear glass, HS-Crimp Neck, rounded bottom (Perkin Elmer)(also in amber glass available) |
20 | 75.5x23 | 100 | 3205121 | |||
Clear glass, HS-Crimp Neck, rounde bottom, with label and graduation lines (Perkin Elmer) | 20 | 75.5x23 | 100 | 3205548 | |||
Clear glass, DIN-Crimp Neck, flat bottom (Carlo Erba, Dani, Fisons, Agilent) |
20 | 75.5x22.5 | 100 | 3205551 | |||
Clear glass, DIN-Crimp Neck, rounded bottom |
20 | 75.5x22.5 | 100 | 3205092 | |||
Amber glass, DIN-Crimp Neck, rounded bottom |
20 | 75.5x22.5 | 100 | 3146114 | |||
Clear glass, DIN-Crimp Neck, flat bottom (Varian) | 5 | 38x20 | 100 | 3205552 | |||
Clear glass, DIN-Crimp Neck, flat bottom (Varian) | 10 | 54.5x23 | 100 | 3205553 |
Other Fisherbrand® Crimp Neck Vials ND20 | |||||||
Type | Capacity ml |
Size mm |
PU / pcs | Item No. | EUR | ||
Clear glass, DIN-Crimp Neck, flat bottom | 50 | 101x31 | 100 | 3205107 | Price Info | ||
Clear glass, DIN-Crimp Neck, flat bottom | 100 | 94.5x51.6 | 88 | 3205547 |
Fisherbrand® Aluminium
Crimp Caps 20mm, with Septa
|
![]() |
Fisherbrand® Aluminium Crimp Caps 20mm, with Septa | |||||||
Type |
Thickness |
Hardness | Cap Design | PU / pcs | Item No. | EUR | |
Butyl, dark grey | 3.0 | 55° shore A | Std. Centre Hole Cap | 100 | 3205094 | Price Info | |
Butyl / PTFE | 3.0 | 50° shore A | Std. Centre Hole Cap | 100 | 3205123 | ||
Pharma-Fix-Septa Butyl / PTFE | 3.0 | 55° shore A | Std. Centre Hole Cap | 100 | 3205531 | ||
Pharma-Fix-Septa, Silicone blue / PTFE grey, UltraClean™ | 3.0 | 45° shore A | Std. Centre Hole Cap | 100 | 3205544 | ||
Silicone blue transparent / PTFE white, UltraClean™ | 3.0 | 45° shore A | Std. Centre Hole Cap | 100 | 3205122 | ||
Silicone white / PTFE beige (Agilent HT Quality) |
3.25 | 45° shore A | Std. Centre Hole Cap | 100 | 3205539 | ||
Silicone white / Aluminium foil silver | 3.0 | 50° shore A | Std. Centre Hole Cap | 100 | 3205537 | ||
Butyl, dark grey | 3.0 | 55° shore A | Headspace Cap | 100 | 3205533 | ||
Butyl / PTFE | 3.0 | 50° shore A | Headspace Cap | 100 | 3205532 | ||
Pharma-Fix-Septa Butyl / PTFE | 3.0 | 50° shore A | Headspace Cap | 100 | 3205535 | ||
Pharma-Fix-Septa, Silicone blue / PTFE grey, UltraClean™ | 3.0 | 45° shore A | Headspace Cap | 100 | 3205545 | ||
Silicone blue transparent / PTFE white, UltraClean™ | 3.0 | 45° shore A | Headspace Cap | 100 | 3205534 | ||
Silicone white / PTFE beige (Agilent Qualität) |
3.25 | 45° shore A | Headspace Cap | 100 | 3205538 | ||
Silicone white / Aluminium foil silver | 3.0 | 50° shore A | Headspace Cap | 100 | 3205536 |
![]() |
Fisherbrand® Magnetic Crimp Neck Caps
20mm,
|
Fisherbrand® Magnetic Crimp Neck Caps 20mm, with Septa, 8mm Hole | |||||||
Type | Thickness mm |
Hardness | PU / pcs | Item No. | EUR | ||
Butyl, dark grey | 3.0 | 55° shore A | 100 | 3205541 | Price Info | ||
Butyl / PTFE | 3.0 | 50° shore A | 100 | 3205542 | |||
Pharma-Fix-Septa Butyl / PTFE | 3.0 | 50° shore A | 100 | 3205543 | |||
Pharma-Fix-Septa, Silicone blue / PTFE grey, UltraClean™ | 3.0 | 45° shore A | 100 | 3205546 | |||
Pharma-Fix-Septa, Silicone blue / PTFE grey, UltraClean™ (Bimetal-Cap) | 3.0 | 45° shore A | 100 | 3205579 | |||
Pharma-Fix-Septa Silicone blue transparent / PTFE white | 3.0 | 45° shore A | 100 | 3205540 |
Other Fisherbrand® Seals 20mm Further seals also with Centre Tear-Off Caps and Complete Tear-Off Caps upon request available. |
Other Fisherbrand® Seals 20mm | |||||||
Type | PU / pcs | Item No. | EUR | ||||
Crimp Neck Aluminium 20mm, hole | 1000 | 3205577 | Price Info | ||||
20mm Butyl injection stopper, grey, only
unassembled available (recommended by Perkin-Elmer) |
100 | 3205124 |
Handheld Crimping Tools |
![]() |
Handheld Crimping Tools | |||||||
Type | PU / pcs | Item No. | EUR | ||||
Crimper for 8mm Crimp Caps | 1 | 3205144 | Price Info | ||||
Crimper for 11mm Crimp Caps | 1 | 3124601 | |||||
Crimper for 13mm Crimp Caps | 1 | 3205495 | |||||
Crimper for 20mm Crimp Caps | 1 | 3205581 | |||||
Decapper for 8mm Crimp Caps | 1 | 3205145 | |||||
Decapper for 11mm Crimp Caps | 1 | 3205582 | |||||
Decapper for 13mm Crimp Caps | 1 | 3205497 | |||||
Decapper for 20mm Crimp Caps | 1 | 3205519 |